Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 206D]

Supplier: Biolegend
320105 320106
320105-BLEA 200.81 USD
320105-BL 320106-BL
Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 206D]
Antibodies
FITC anti-human FOXP3 [206D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Pigtailed Macaque; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 206D
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Pigtailed Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR