Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 561]

Supplier: Biolegend
343603 343604
343603-BLEA 113.7 USD
343603-BL 343604-BL
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 561]
Antibodies
FITC anti-human CD34 [561]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 561
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR