Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 581]

Supplier: Biolegend
343523 343524
343523-BLEA 114.3 USD
343523-BL 343524-BL
Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 581]
Antibodies
Biotin anti-human CD34 [581]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR