Anti-C3AR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: hC3aRZ8]

Supplier: Biolegend
345805 345806
345805-BLEA 173.4 USD
345805-BL 345806-BL
Anti-C3AR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: hC3aRZ8]
Antibodies
APC anti-human C3AR [hC3aRZ8]; Isotype: Mouse IgG2b; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: C3AR
Clonality: Monoclonal
Clone: hC3aRZ8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR