Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20049 20049-1
71004-150EA 217.08 USD
71004-150 71004-152
Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR