Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MPC-11]

Supplier: Biolegend
400301 400302
400301-BLEA 35.28 USD
400301-BL 400302-BL
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody [clone: MPC-11]
Antibodies
Purified Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC, ICC, IF, IHC, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 95% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR