Anti-IL-23 (p19) Mouse Monoclonal Antibody [clone: HLT2736]

Supplier: Biolegend
511201 511202
511201-BLEA 132.9 USD
511201-BL 511202-BL
Anti-IL-23 (p19) Mouse Monoclonal Antibody [clone: HLT2736]
Antibodies
Purified anti-human IL-23 (p19) [HLT2736]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-23 p19, does not cross react with mouse.; Apps: WB, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting applications, a concentration range of 0.5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-23 (p19)
Clonality: Monoclonal
Clone: HLT2736
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;IL-23 p19;does not cross react with mouse.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR