Anti-IL-1b Mouse Monoclonal Antibody [clone: JK1B-1]

Supplier: Biolegend
508201 508202
508201-BLEA 119.99 USD
508201-BL 508202-BL
Anti-IL-1b Mouse Monoclonal Antibody [clone: JK1B-1]
Antibodies
Purified anti-human IL-1β [JK1B-1]; Isotype: Mouse IgG; Reactivity: Human; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-1β recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-1b
Clonality: Monoclonal
Clone: JK1B-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR