Anti-IgG2a, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-173]

Supplier: Biolegend
400211 400212
400211-BLEA 50.7 USD
400211-BL 400212-BL
Anti-IgG2a, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-173]
Antibodies
PE Mouse IgG2a, κ Isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR