Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-21]

Supplier: Biolegend
400111 400112
400111-BLEA 45.3 USD
400111-BL 400112-BL
Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MOPC-21]
Antibodies
PE Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg

Each lot of this mouse IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR