Anti-ICOS (CD278) Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15F9]

Supplier: Biolegend
107705 107706
107705-BLEA 137.3 USD
107705-BL 107706-BL
Anti-ICOS (CD278) Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15F9]
Antibodies
PE anti-mouse CD278 (ICOS) [15F9]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: ICOS (CD278)
Clonality: Monoclonal
Clone: 15F9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR