Anti-CD278 (ICOS) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7E.17G9]

Supplier: Biolegend
117405 117406
117405-BLEA 137.69 USD
117405-BL 117406-BL
Anti-CD278 (ICOS) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7E.17G9]
Antibodies
PE anti-mouse CD278 (ICOS) [7E.17G9]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: 7E.17G9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR