Anti-CD19 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 6D5]

Supplier: Biolegend
115523 115526
115523-BLEA 253.11 USD
115523-BL 115526-BL
Anti-CD19 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 6D5]
Antibodies
Pacific Blue™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR