Anti-CD19 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIB19]

Supplier: Biolegend
302223 302224 302232
302223-BLEA 145.89 USD
302223-BL 302224-BL 302232-BL
Anti-CD19 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIB19]
Antibodies
Pacific Blue™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test sizes, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume. 
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 0.5 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR