Anti-TCR Vb7 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TR310]

Supplier: Biolegend
118305 118306
118305-BLEA 105.8 USD
118305-BL 118306-BL
Anti-TCR Vb7 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TR310]
Antibodies
FITC anti-mouse TCR Vβ7 [TR310]; Isotype: Rat IgG2b, κ; Reactivity: Mouse TCR Vβ7 bearing T cells and NKT cells; may not be present in mice expressing the MIS-1a haplotype; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR Vb7
Clonality: Monoclonal
Clone: TR310
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse TCR Vβ7 bearing T cells and NKT cells; may not be present in mice expressing the MIS-1a haplotype
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR