Anti-IgMb Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AF6-78]

Supplier: Biolegend
406205 406206
406205-BLEA 119.4 USD
406205-BL 406206-BL
Anti-IgMb Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AF6-78]
Antibodies
FITC anti-mouse IgMb [AF6-78]; Isotype: Mouse (BALB/c) IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgMb
Clonality: Monoclonal
Clone: AF6-78
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR