Anti-CD4 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM4-4]

Supplier: Biolegend
116003 116004
116003-BLEA 105.8 USD
116003-BL 116004-BL
Anti-CD4 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM4-4]
Antibodies
FITC anti-mouse CD4 [RM4-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR