Anti-TCR Vd2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B6]

Supplier: Biolegend
331405 331406
331405-BLEA 105.8 USD
331405-BL 331406-BL
Anti-TCR Vd2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B6]
Antibodies
FITC anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactive: Chimpanzee, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: TCR Vd2
Clonality: Monoclonal
Clone: B6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-reactive: Chimpanzee;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR