Anti-CD44 Rat Monoclonal Antibody (Biotin) [clone: IM7]

Supplier: Biolegend
103003 103004
103003-BLEA 47.3 USD
103003-BL 103004-BL
Anti-CD44 Rat Monoclonal Antibody (Biotin) [clone: IM7]
Antibodies
Biotin anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Horse (Equine), Cattle (Bovine, Cow), Swine (Pig, Porcine), Dog (Canine), Cat (Feline); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Squirrel Monkey;Horse (Equine);Cattle (Bovine;Cow);Swine (Pig;Porcine);Dog (Canine);Cat (Feline)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR