Anti-H-2Kb Mouse Monoclonal Antibody (Biotin) [clone: AF6-88.5]

Supplier: Biolegend
116503 116504
116503-BLEA 93.4 USD
116503-BL 116504-BL
Anti-H-2Kb Mouse Monoclonal Antibody (Biotin) [clone: AF6-88.5]
Antibodies
Biotin anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone: AF6-88.5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse b haplotype;Does not cross-react with other haplotypes (e.g.;d;f;j;k;p;q;r;s;u;v)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR