Anti-IgG2a, k Isotype Ctrl Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MOPC-173]

Supplier: Biolegend
400221 400222
400221-BLEA 151.3 USD
400221-BL 400222-BL
Anti-IgG2a, k Isotype Ctrl Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MOPC-173]
Antibodies
APC Mouse IgG2a, κ Isotype Ctrl (FC) [MOPC-173]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 tests

Each lot of this mouse IgG2a, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR