Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MOPC-21]

Supplier: Biolegend
400119 400120
400119-BLEA 45.3 USD
400119-BL 400120-BL
Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MOPC-21]
Antibodies
APC Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR