Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6D5]

Supplier: Biolegend
115511 115512
115511-BLEA 58.9 USD
115511-BL 115512-BL
Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6D5]
Antibodies
APC anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR