Anti-CD19 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 6D5]

Supplier: Biolegend
115527 115528
115527-BLEA 110.7 USD
115527-BL 115528-BL
Anti-CD19 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 6D5]
Antibodies
Alexa Fluor® 700 anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR