Anti-CD29 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: TS2/16]

Supplier: Biolegend
303019 303020
303019-BLEA 139.6 USD
303019-BL 303020-BL
Anti-CD29 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: TS2/16]
Antibodies
Alexa Fluor® 700 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD29
Clonality: Monoclonal
Clone: TS2/16
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Cynomolgus;Dog (Canine);(Horse reactivity);Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR