Anti-IgD Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 11-26c.2a]

Supplier: Biolegend
405707 405708
405707-BLEA 126.1 USD
405707-BL 405708-BL
Anti-IgD Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 11-26c.2a]
Antibodies
Alexa Fluor® 647 anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgD
Clonality: Monoclonal
Clone: 11-26c.2a
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR