Anti-CD49d Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: R1-2]

Supplier: Biolegend
103613 103614
103613-BLEA 113.7 USD
103613-BL 103614-BL
Anti-CD49d Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: R1-2]
Antibodies
Alexa Fluor® 647 anti-mouse CD49d [R1-2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: R1-2
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR