Anti-TRA-1-81 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TRA-1-81]

Supplier: Biolegend
330705 330706
330705-BLEA 185.05 USD
330705-BL 330706-BL
Anti-TRA-1-81 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TRA-1-81]
Antibodies
Alexa Fluor® 647 anti-human TRA-1-81 [TRA-1-81]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TRA-1-81
Clonality: Monoclonal
Clone: TRA-1-81
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR