Anti-CD29 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TS2/16]

Supplier: Biolegend
303017 303018
303017-BLEA 140.28 USD
303017-BL 303018-BL
Anti-CD29 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TS2/16]
Antibodies
Alexa Fluor® 647 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD29
Clonality: Monoclonal
Clone: TS2/16
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Cynomolgus;Dog (Canine);(Horse reactivity);Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR