Anti-I-Ad Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 39-10-8]

Supplier: BioLegend
115008 115008
115008-BLEA 254.58 USD
115008-BL CA115008-BL
Anti-I-Ad Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 39-10-8]
Antibodies
Alexa Fluor® 488 anti-mouse I-Ad [39-10-8]; Isotype: Mouse (C3H.SW) IgG3, κ; Reactivity: Mouse d haplotype, Does not cross-react with other haplotypes (e.g., a, b, k, p, q, s); Apps: FC, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-Ad
Clonality: Monoclonal
Clone: 39-10-8
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse (C3H.SW) IgG3, κ
Reactivity: Mouse d haplotype;Does not cross-react with other haplotypes (e.g.;a;b;k;p;q;s)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR