CT-600 Series Constant Temperature Bath, 115 V, Cannon Instrument Company
Supplier: Cannon Instrument Company
9726-A36
10791-426EA 13536.29 USD
10791-426
CT-600 Series Constant Temperature Bath, 115 V, Cannon Instrument Company
Baths Viscometer Baths
The CT-600 is similar to the CT-500 but includes built-in thermoelectric cooling modules, making it ideal for controlled temperature applications between 10°C and 30°C
The CT-600 provide accurate temperature control per ASTM D 445 specifications between 10°C and 100°C.