CT-1000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Supplier: Cannon Instrument Company
9726-A20
47735-066EA 14190.64 USD
47735-066
CT-1000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Baths Viscometer Baths
This durable and accurate insulated bath offers quick-set temperature control options and ASTM D 445 reliability to 150°C
Up to seven viscometer tubes/holders can be placed in the bath. The unit is equipped with several safety features, including a bath level float and over-temperature thermistor.