Trixylenyl phosphate

Supplier: Pfaltz & Bauer
Danger

Synonyms: XylylphosphateAntiblaze TXPTXP

T32575-100G
101178-512EA 356.22 USD
101178-512
Trixylenyl phosphate
Trixylenyl phosphate
Formula: C₂₄H₂₇O₄P
MW: 202.144301 g/mol
Boiling Pt: 490…491 °C
Density: 1.136
Flash Pt: 232 °C
MDL Number: MFCD01778964
CAS Number: 25155-23-1
EINECS: 246-677-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR