Chromocult® Enterococci Broth, MilliporeSigma
Supplier: MilliporeSigma
1.10294.0500
EM1.10294.0500EA 473.65 USD
EM1.10294.0500
Chromocult® Enterococci Broth, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
  • Fair to very good growth in Enterococcus faecalis ATCC 11700
  • Fair to very good growth in Enterococcus faecalis ATCC 19433 (WDCM 00009)
  • Fair to very good growth in Enterococcus faecium ATCC 6057
  • Fair to very good growth in Enterococcus durans ATCC 19432
  • No growth to very good growth in Staphylococcus aureus ATCC 25923 (WDCM 00034)
  • No growth to poor growth in Aeromonas hydrophila ATCC 7966 (WDCM 00063)
  • No growth to poor growth in Escherichia coli ATCC 25922 (WDCM 00013)
  • No growth to poor growth in Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)
  • + color change to blue-green in Enterococcus faecalis ATCC 11700
  • + color change to blue-green in Enterococcus faecalis ATCC 19433 (WDCM 00009)
  • + color change to blue-green in Enterococcus faecium ATCC 6057
  • + color change to blue-green in Enterococcus durans ATCC 19432
  • - color change to blue-green in Staphylococcus aureus ATCC 25923 (WDCM 00034)
  • - color change to blue-green in Escherichia coli ATCC 25922 (WDCM 00013)
  • - color change to blue-green in Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)