Plate Count Agar, MilliporeSigma
Supplier: MilliporeSigma
1.05463.5000
EM1.05463.5000EA 1436.63 USD
EM1.05463.5000
Plate Count Agar, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Casein-peptone glucose yeast extract agar for microbiology. Peptone from casein 5.0; Yeast extract 2.5; D(+)Glucose 1.0; Agar-agar 14.0.
  • 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
  • 10 to 100 inoculum on reference medium Escherichia coli ATCC 25922 (WDCM 00013)
  • 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
  • 10 to 100 inoculum on reference medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
  • 10 to 100 inoculum on reference medium Listeria monocytogenes ATCC 19118
  • 10 to 100 inoculum on reference medium Lactobacillus acidophilus ATCC 4356 (WDCM 00098)
  • 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
  • 10 to 100 inoculum on reference medium Escherichia coli ATCC 25922 (WDCM 00013)
  • 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 8739 (WDCM 00012)
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Greater than or equal to 70% recovery on test medium Bacillus subtilis ATCC 6633 (WDCM 00003)
  • Greater than or equal to 70% recovery on test medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
  • Greater than or equal to 70% recovery on test medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
  • Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 19118
  • Greater than or equal to 70% recovery on test medium Lactobacillus acidophilus ATCC 4356 (WDCM 00098)
  • Greater than or equal to 70% recovery on test medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 8739 (WDCM 00012)
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Greater than or equal to 70% recovery on test medium Bacillus subtilis ATCC 6633 (WDCM 00003)