MacCONKEY Agar, MilliporeSigma
Supplier: MilliporeSigma
1.00205.5000
EM1.00205.5000EA 2033.67 USD
EM1.00205.5000
MacCONKEY Agar, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Peptone from gelatin 17.0; peptone from casein 1.5; peptone from meat 1.5; sodium chloride 5.0; lactose 10.0; bile salt mixture 1.5; neutral red 0.03; crystal violet 0.001; agar-agar 13.5.
In North America 1.00205.0500 replaces and is identical to 1.00205.5007