Anti-IgY Rabbit Antibody (CF™ 594)

Supplier: Biotium

20164-1MG
89493-288EA 222.25 USD
89493-288
Anti-IgY Rabbit Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgY
Clonality:
Clone:
Conjugation: CF™ 594
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR