Anti-IgY Goat Antibody (CF™ 568)

Supplier: Biotium

20104-1MG
89493-194EA 266.7 USD
89493-194
Anti-IgY Goat Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgY
Clonality:
Clone:
Conjugation: CF™ 568
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR