Anti-IgY Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20311-1MG
89493-440EA 266.7 USD
89493-440
Anti-IgY Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR