Anti-53BP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70316
89337-300EA 600.98 USD
89337-300
Anti-53BP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to human 53BP1 ( amino terminus )

For WB: Use at a dilution of 1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: 53BP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR