Anti-C6ORF10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX46401
89269-078EA 630.56 USD
89269-078
Anti-C6ORF10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C6ORF10 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: C6ORF10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR