Anti-FKBP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX44851
89266-196EA 630.56 USD
89266-196
Anti-FKBP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FKBP2 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: FKBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR