Thioacetamide ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethanethioamide

225895-100G 225895-25G 225895-500G
BT225895-100GEA 58.47 USD
BT225895-100G BT225895-25G BT225895-500G
Thioacetamide ≥98%
Thioacetamide

CAS: 62-55-5; EC No: 200-541-4; MDL No: MFCD00008070; RTECS: AC8925000 Crystalline; Linear Formula: CH3CSNH2; MW: 75.13 Melting Point: 108-112°


  • RTECS: AC8925000
  • Crystalline

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 108…112 °C
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR