Potassium iodide ≥99%

Supplier: BeanTown Chemical

225810-1KG 225810-250G 225810-50G
BT225810-1KGEA 265.78 USD
BT225810-1KG BT225810-250G BT225810-50G
Potassium iodide ≥99%
Potassium iodide

CAS: 7681-11-0; EC No: 231-659-4; MDL No: MFCD00011405; RTECS: TT2975000 Powder; Linear Formula: KI; MW: 166.01 Melting Point: 681°; Boiling Point: 1330° Density (g/mL): 3.13; Refractive Index: 1.677 Air Sensitive, Light Sensitive, Moisture Sensitive


  • RTECS: TT2975000
  • Refractive index: 1.677
  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Powder

Formula: Kl
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.13 g/mL
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR