Silica gel, Pore size: 60 Å 0.075-0.2mm (70-200 mesh)
Supplier: BeanTown Chemical
225695-1KG
225695-250G
225695-50G
BT225695-1KGEA
194.2
USD
BT225695-1KG
BT225695-250G
BT225695-50G
Silica gel, Pore size: 60 Å 0.075-0.2mm (70-200 mesh)
Silica gel
CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232
Powder, S.A. 470-570 m2/g
Melting Point:
- Hygroscopic
- Powder, S.A. 470-570 m²/g
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: >1600 °C |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...