Poly(vinylidene fluoride)

Supplier: BeanTown Chemical

225570-2KG 225570-500G
BT225570-2KGEA 303.8 USD
BT225570-2KG BT225570-500G
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)

CAS: 24937-79-9; MDL No: MFCD00084470 Powder; Linear Formula: (-CH2CF2-)n Melting Point: 155-160°

Formula: (-CH₂CF₂-)n
Melting Pt: 150…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00084470
CAS Number: 24937-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR