Lactose, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Milk sugarD-LactoseAnhydrous Lactose(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal

225065-500G 225065-1KG
BT225065-1KGEA 175.07 USD
BT225065-1KG BT225065-500G
Lactose, anhydrous ≥98%
Lactose

  • RTECS: OD9625000
  • Powder

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR