Polyethylene glycol 200

Supplier: BeanTown Chemical

Synonyms: Ethylene glycol polymer average Mn 200PEG 200

225040-1KG 225040-500G
BT225040-500GEA 56.88 USD
BT225040-500G BT225040-1KG
Polyethylene glycol 200
Polyethylene glycol 200

  • RTECS: TQ3600000
  • Refractive index: 1.459
  • Hygroscopic
  • Liquid

Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
Melting Pt: –65 °C
Density: 1.127 g/mL
Flash Pt: 171 °C (339 °F)
MDL Number: MFCD01779596
CAS Number: 25322-68-3
EINECS: 203-473-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR