Silver ≥99.9% (trace metals basis), wire Ø 0.25 mm, hard type

Supplier: BeanTown Chemical

224900-100M 224900-25M 224900-500M
BT224900-100MEA 274 USD
BT224900-100M BT224900-25M BT224900-500M
Silver ≥99.9% (trace metals basis), wire Ø 0.25 mm, hard type
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397 Solid; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • Solid

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR