Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 2,000

Supplier: BeanTown Chemical

224850-1L 224850-250ML 224850-50ML
BT224850-1LEA 121 USD
BT224850-1L BT224850-250ML BT224850-50ML
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 2,000
Polydimethylsiloxane, trimethylsiloxy terminated

CAS: 9016-00-6; MDL No: MFCD00148360; RTECS: TQ2690000 Liquid; Linear Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 ; MW ≈2,000 Melting Point: -40°; Boiling Point: 205°; Flash point: 232°C (450°F) Density (g/mL): 0.966; Refractive Index: 1.400


  • Refractive Index: 1.400

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Boiling Pt: >205 °C
Melting Pt: –40 °C
Density: 0.966 g/ml
Flash Pt: 232 °C
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR